accessibility menu, dialog, popup

UserName

CH3CH2

CAS RN 155041-85-3

IUPAC Name: 4-ethenyl-4'-(4-methylphenyl)-1,1'-bi(cyclohexane)
Synonyms: 4-ethenyl-4'-(4-methylphenyl)-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 282.47
Molecular Formula: C21H30
InChi Key: BCUMFFLWJNKNOU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1CCC(CC1)C1CCC(CC1)C=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results