missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-propylbicyclohexyl 98.0+%, TCI America™

Catalog No. p-7129368
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11571G 1 g
E11575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11571G Supplier TCI America Supplier No. E11571G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123560-48-5
Molecular Formula C23H34F2O
Molecular Weight (g/mol) 364.521
MDL Number MFCD18433524
InChI Key CFJCNWQXOICKIF-UHFFFAOYSA-N
Synonym trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, trans,trans-4'-4-ethoxy-2,3-difluoro-phenyl-4-propyl-bicyclohexyl, trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-n-propylbicyclohexyl, 1r,1'r,4r,4's-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1-ethoxy-2,3-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-4-ethoxy-2,3-difluorophenyl-4'-propylbicyclohexyl, trans-4-4-ethoxy-2,3-difluorophenyl-trans-4'-propylbicyclohexane, benzene, 1-ethoxy-2,3-difluoro-4-4'-propyl 1,1'-bicyclohexyl-4-yl, trans-4-4-ethoxy-2,3-difluoro-phenyl-4-propyl-bicycloh, 1r,1's,4r,4'r-4-4-ethoxy-2,3-difluorophenyl-4'-propyl-1,1'-bi cyclohexane
PubChem CID 11417199
IUPAC Name 1-ethoxy-2,3-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene
SMILES CCCC1CCC(CC1)C2CCC(CC2)C3=C(C(=C(C=C3)OCC)F)F

Specifications

Melting Point 79°C
Color White
Quantity 1 g
Formula Weight 364.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4′-propylbicyclohexyl