missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4'-(3-Butenyl)-4-(p-tolyl)bicyclohexyl 98.0+%, TCI America™

Catalog No. B49151G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49151G 1 g
B49155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49151G Supplier TCI America Supplier No. B49151G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 129738-42-7
Molecular Formula C23H34
Molecular Weight (g/mol) 310.525
MDL Number MFCD13188633
InChI Key OXPUOKDPOMJNKA-UHFFFAOYSA-N
PubChem CID 18724321
IUPAC Name 1-[4-(4-but-3-enylcyclohexyl)cyclohexyl]-4-methylbenzene
SMILES CC1=CC=C(C=C1)C2CCC(CC2)C3CCC(CC3)CCC=C

Specifications

Melting Point 103°C
Color White
Quantity 1 g
Synonym trans,trans-4′C-(3-Butenyl)-4-(4-methylphenyl)bicyclohexyl
Formula Weight 310.53
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4′-(3-Butenyl)-4-(p-tolyl)bicyclohexyl
EINECSNumber (4)-1882
TSCA No