accessibility menu, dialog, popup

UserName

CH3CH2

CAS RN 129738-42-7

IUPAC Name: 4-(but-3-en-1-yl)-4'-(4-methylphenyl)-1,1'-bi(cyclohexane)
Synonyms: 4-(but-3-en-1-yl)-4'-(4-methylphenyl)-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 310.53
Molecular Formula: C23H34
InChi Key: OXPUOKDPOMJNKA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1CCC(CC1)C1CCC(CCC=C)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results