missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl 98.0+%, TCI America™

Catalog No. D48721G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48721G 1 g
D48725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48721G Supplier TCI America Supplier No. D48721G

Chemical Identifiers

CAS 142400-92-8
Molecular Formula C20H26F2
Molecular Weight (g/mol) 304.425
MDL Number MFCD09842838
InChI Key ALFLDQIYGBNZCO-UHFFFAOYSA-N
Synonym trans,trans-4-3,4-difluorophenyl-4'-vinyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-vinylbicyclohexyl, trans-4-3,4-difluorophenyl-trans-4'-vinylbicyclohexane, benzene, 4-trans,trans-4'-ethenyl 1,1'-bicyclohexyl-4-yl-1,2-difluoro, trans trans-4-4'-ethenyl 1,1'-bicyclohexyl-4-yl-1,2-difluorobenzene, trans,trans-4-3,4-difluoro-phenyl-4'-vinyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-vinylbi cyclohexane, 4-e-1,2-dicyclohexylethenyl-1,2-difluorobenzene, 4-vinyl-4'-3,4-difluorophenyl-1,1'-bicyclohexane, trans-4-3,4-difluorophenyl-trans-4-vinylbicyclohexane
PubChem CID 12010662
IUPAC Name 4-[4-(4-ethenylcyclohexyl)cyclohexyl]-1,2-difluorobenzene
SMILES C=CC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F

Specifications

Melting Point 46°C
Color White
Quantity 1 g
Formula Weight 304.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4-(3,4-Difluorophenyl)-4′-vinylbicyclohexyl