missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-4-(3,4-Difluorophenyl)-4'-propylbicyclohexyl 98.0+%, TCI America™
$72.70 - $255.38
Chemical Identifiers
| CAS | 82832-57-3 |
|---|---|
| Molecular Formula | C21H30F2 |
| Molecular Weight (g/mol) | 320.468 |
| MDL Number | MFCD09838997 |
| InChI Key | FSWZOZXLWVWJAH-UHFFFAOYSA-N |
| Synonym | trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl |
| PubChem CID | 581325 |
| IUPAC Name | 1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene |
| SMILES | CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F |
Chemical Identifiers
| 82832-57-3 | |
| 320.468 | |
| FSWZOZXLWVWJAH-UHFFFAOYSA-N | |
| 581325 | |
| CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F |
| C21H30F2 | |
| MFCD09838997 | |
| trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl | |
| 1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene |
Specifications
| 82832-57-3 | |
| White | |
| MFCD09838997 | |
| trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl | |
| CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F | |
| 320.468 | |
| 320.47 | |
| Crystalline Powder |
| 44°C | |
| C21H30F2 | |
| 1 g | |
| FSWZOZXLWVWJAH-UHFFFAOYSA-N | |
| 1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene | |
| 581325 | |
| ≥98.0% (GC) | |
| trans,trans-4-(3,4-Difluorophenyl)-4′-propylbicyclohexyl |
Safety and Handling
EINECSNumber : (4)-1883
TSCA : No