missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-(3,4-Difluorophenyl)-4'-pentylbicyclohexyl 98.0+%, TCI America™

Catalog No. D47985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47985G 5 g
D47981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47985G Supplier TCI America Supplier No. D47985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118164-51-5
Molecular Formula C23H34F2
Molecular Weight (g/mol) 348.522
MDL Number MFCD13182310
InChI Key YDVUSMRUBCJGAV-UHFFFAOYSA-N
Synonym trans,trans-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-pentylbicyclohexyl, 4-trans-4-trans-4-pentylcyclohexyl cyclohexyl-1,2-difluorobenzene, benzene, 1,2-difluoro-4-4'-pentyl 1,1'-bicyclohexyl-4-yl, 1,2-difluoro-4-trans,trans-4'-pentyl 1,1'-bicyclohexyl-4-yl benzene, 1,2-difluoro-4-4-4-pentylcyclohexyl cyclohexyl benzene, 4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-n-pentylbicyclohexyl, 1r,1's,4r,4'r-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-pentylbicyclohexyl-3,4-difluorobenzene
PubChem CID 581521
IUPAC Name 1,2-difluoro-4-[4-(4-pentylcyclohexyl)cyclohexyl]benzene
SMILES CCCCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F

Specifications

Melting Point 48°C
Color White
Quantity 5 g
Formula Weight 348.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4-(3,4-Difluorophenyl)-4′-pentylbicyclohexyl
EINECSNumber (4)-1855
TSCA No