missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl, 97%

CAS: 82832-57-3 | C21H30F2 | 320.468 g/mol

$74.32 - $248.90

Chemical Identifiers

CAS 82832-57-3
Molecular Formula C21H30F2
Molecular Weight (g/mol) 320.468
MDL Number MFCD09838997
InChI Key FSWZOZXLWVWJAH-UHFFFAOYSA-N
Synonym trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl
PubChem CID 581325
IUPAC Name 1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene
SMILES CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5535603
View Documents
Thermo Scientific Chemicals
H5535603
1 g
Each for $74.32
Only null left
 
AAH5535606
View Documents
Thermo Scientific Chemicals
H5535606
5 g
Each for $248.90
Only null left
 
Description

Description

It is  used in liquid crystal monomer, pharmaceutical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is  used in liquid crystal monomer, pharmaceutical synthesis.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications

Chemical Identifiers

82832-57-3
320.468
FSWZOZXLWVWJAH-UHFFFAOYSA-N
581325
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F
C21H30F2
MFCD09838997
trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl
1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene

Specifications

82832-57-3
C21H30F2
1 g
Slightly soluble in water.
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F
320.468
320.47
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl
43°C to 46°C
MFCD09838997
trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl
FSWZOZXLWVWJAH-UHFFFAOYSA-N
1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene
581325
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only