missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-3,4,5-Trifluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl 98.0+%, TCI America™

Catalog No. p-7137694
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33191G 1 g
T33195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33191G Supplier TCI America Supplier No. T33191G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137529-41-0
Molecular Formula C27H33F3
Molecular Weight (g/mol) 414.56
MDL Number MFCD13188632
InChI Key LQRASBADJFXRBP-UHFFFAOYSA-N
PubChem CID 22373586
IUPAC Name 3,4,5-trifluoro-4'-{4'-propyl-[1,1'-bi(cyclohexane)]-4-yl}-1,1'-biphenyl
SMILES CCCC1CCC(CC1)C1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C(F)=C1

Specifications

Melting Point 103°C
Color White
Quantity 1 g
Formula Weight 414.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-3,4,5-Trifluoro-4′-(4′-propylbicyclohexyl-4-yl)biphenyl