missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one 98.0+%, TCI America™

Catalog No. B4468200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4468200MG 200 mg
B44681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4468200MG Supplier TCI America Supplier No. B4468200MG
Only null left

Chemical Identifiers

CAS 103836-71-1
Molecular Formula C19H12F6O
Molecular Weight (g/mol) 370.294
InChI Key OIUBVYQZMUKRRF-YDWXAUTNSA-N
Synonym trans,trans-Bis[4-(trifluoromethyl)benzal]acetone, trans,trans-Bis[4-(trifluoromethyl)benzylidene]acetone, trans,trans-Bis[4-(trifluoromethyl)styryl] Ketone
PubChem CID 5373276
IUPAC Name (1E,4E)-1,5-bis[4-(trifluoromethyl)phenyl]penta-1,4-dien-3-one
SMILES C1=CC(=CC=C1C=CC(=O)C=CC2=CC=C(C=C2)C(F)(F)F)C(F)(F)F

Specifications

Quantity 200 mg
Formula Weight 370.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one