missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one 98.0+%, TCI America™
$98.79 - $331.81
Chemical Identifiers
| CAS | 37951-12-5 |
|---|---|
| Molecular Formula | C19H18O3 |
| Molecular Weight (g/mol) | 294.35 |
| MDL Number | MFCD00014901 |
| InChI Key | IOZVKDXPBWBUKY-LQIBPGRFSA-N |
| Synonym | trans,trans-Bis(4-methoxybenzal)acetone, trans,trans-Bis(4-methoxybenzylidene)acetone, trans,trans-Bis(4-methoxystyryl) Ketone |
| PubChem CID | 715840 |
| IUPAC Name | (1E,4E)-1,5-bis(4-methoxyphenyl)penta-1,4-dien-3-one |
| SMILES | COC1=CC=C(C=C1)C=CC(=O)C=CC2=CC=C(C=C2)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4467200MG
|
TCI America
B4467200MG |
200 mg |
Each for $98.79
|
|
|||||
|
B44671G
|
TCI America
B44671G |
1 g |
Each for $331.81
|
|
|||||
Chemical Identifiers
| 37951-12-5 | |
| 294.35 | |
| IOZVKDXPBWBUKY-LQIBPGRFSA-N | |
| 715840 | |
| COC1=CC=C(C=C1)C=CC(=O)C=CC2=CC=C(C=C2)OC |
| C19H18O3 | |
| MFCD00014901 | |
| trans,trans-Bis(4-methoxybenzal)acetone, trans,trans-Bis(4-methoxybenzylidene)acetone, trans,trans-Bis(4-methoxystyryl) Ketone | |
| (1E,4E)-1,5-bis(4-methoxyphenyl)penta-1,4-dien-3-one |
Specifications
| 37951-12-5 | |
| Yellow | |
| MFCD00014901 | |
| trans,trans-Bis(4-methoxybenzal)acetone, trans,trans-Bis(4-methoxybenzylidene)acetone, trans,trans-Bis(4-methoxystyryl) Ketone | |
| COC1=CC=C(C=C1)C=CC(=O)C=CC2=CC=C(C=C2)OC | |
| 294.35 | |
| 294.35 | |
| Crystalline Powder |
| 129°C | |
| C19H18O3 | |
| 200 mg | |
| IOZVKDXPBWBUKY-LQIBPGRFSA-N | |
| (1E,4E)-1,5-bis(4-methoxyphenyl)penta-1,4-dien-3-one | |
| 715840 | |
| ≥98.0% (GC) | |
| trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one |
Safety and Handling
EINECSNumber : (4)-0856
TSCA : No