CAS RN 37951-12-5
IUPAC Name:
1,5-bis(4-methoxyphenyl)penta-1,4-dien-3-one
Synonyms:
1,5-bis(4-methoxyphenyl)penta-1,4-dien-3-one
Molecular Weight (g/mol):
294.35
Molecular Formula:
C19H18O3
InChi Key:
IOZVKDXPBWBUKY-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=CC(=O)C=CC2=CC=C(OC)C=C2)C=C1
1
–
1
of
1
results
1
trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one 98.0+%, TCI America™
CAS: 37951-12-5 Molecular Formula: C19H18O3 Molecular Weight (g/mol): 294.35 MDL Number: MFCD00014901 InChI Key: IOZVKDXPBWBUKY-LQIBPGRFSA-N Synonym: trans,trans-Bis(4-methoxybenzal)acetone, trans,trans-Bis(4-methoxybenzylidene)acetone, trans,trans-Bis(4-methoxystyryl) Ketone PubChem CID: 715840 IUPAC Name: (1E,4E)-1,5-bis(4-methoxyphenyl)penta-1,4-dien-3-one SMILES: COC1=CC=C(C=C1)C=CC(=O)C=CC2=CC=C(C=C2)OC