missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 129°C |
| Color | Yellow |
| Quantity | 200 mg |
| Formula Weight | 294.35 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one |
Safety and Handling
| EINECSNumber | (4)-0856 |
| TSCA | No |