Learn More
trans,trans-1,4-Diphenyl-1,3-butadiene, 98+%
CAS: 538-81-8 | C16H14 | 206.29 g/mol
Supplier: Thermo Scientific Chemicals A1753122
Description
trans,trans-1,4-Diphenyl-1,3-butadiene reacts with Rieke metal complexes of barium and strontium to prepare metal-diene reagents, which is used for the carbocyclization with dichloroalkanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans,trans-1,4-Diphenyl-1,3-butadiene reacts with Rieke metal complexes of barium and strontium to prepare metal-diene reagents, which is used for the carbocyclization with dichloroalkanes.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| trans,trans-1,4-Diphenyl-1,3-butadiene | |
| 151°C to 154°C | |
| Odorless | |
| MFCD00004791 | |
| 1905937 | |
| bistyryl, 1,4-diphenyl-1,3-butadiene, 1,4-diphenylbutadiene, trans,trans-1,4-diphenyl-1,3-butadiene, 1,4-diphenylerythrene, distyryl, trans,trans-1,4-diphenylbuta-1,3-diene, 1,4-diphenylbuta-1,3-diene, 1,1'-1e,3e-buta-1,3-diene-1,4-diyldibenzene, 1,3-butadiene, 1,4-diphenyl | |
| JFLKFZNIIQFQBS-FNCQTZNRSA-N | |
| [(1E,3E)-4-phenylbuta-1,3-dienyl]benzene | |
| 641683 | |
| 206.29 |
| 538-81-8 | |
| 350°C (720 mmHg) | |
| C16H14 | |
| 100 g | |
| 14,3320 | |
| Slightly soluble in water. | |
| C(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1 | |
| 206.29 | |
| CHEBI:35100 | |
| ≥98% |
Chemical Identifiers
| 538-81-8 | |
| 206.29 | |
| JFLKFZNIIQFQBS-FNCQTZNRSA-N | |
| 641683 | |
| [(1E,3E)-4-phenylbuta-1,3-dienyl]benzene |
| C16H14 | |
| MFCD00004791 | |
| bistyryl, 1,4-diphenyl-1,3-butadiene, 1,4-diphenylbutadiene, trans,trans-1,4-diphenyl-1,3-butadiene, 1,4-diphenylerythrene, distyryl, trans,trans-1,4-diphenylbuta-1,3-diene, 1,4-diphenylbuta-1,3-diene, 1,1'-1e,3e-buta-1,3-diene-1,4-diyldibenzene, 1,3-butadiene, 1,4-diphenyl | |
| CHEBI:35100 | |
| C(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 212-952-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only