missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-1,4-Diphenyl-1,3-butadiene, 98+%

Catalog No. AAA1753106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1753106 5 g
AAA1753114 25 g
AAA1753122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1753106 Supplier Thermo Scientific Chemicals Supplier No. A1753106

CAS: 538-81-8 | C16H14 | 206.29 g/mol

trans,trans-1,4-Diphenyl-1,3-butadiene reacts with Rieke metal complexes of barium and strontium to prepare metal-diene reagents, which is used for the carbocyclization with dichloroalkanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-1,4-Diphenyl-1,3-butadiene reacts with Rieke metal complexes of barium and strontium to prepare metal-diene reagents, which is used for the carbocyclization with dichloroalkanes.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 538-81-8
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00004791
InChI Key JFLKFZNIIQFQBS-FNCQTZNRSA-N
Synonym bistyryl, 1,4-diphenyl-1,3-butadiene, 1,4-diphenylbutadiene, trans,trans-1,4-diphenyl-1,3-butadiene, 1,4-diphenylerythrene, distyryl, trans,trans-1,4-diphenylbuta-1,3-diene, 1,4-diphenylbuta-1,3-diene, 1,1'-1e,3e-buta-1,3-diene-1,4-diyldibenzene, 1,3-butadiene, 1,4-diphenyl
PubChem CID 641683
ChEBI CHEBI:35100
IUPAC Name [(1E,3E)-4-phenylbuta-1,3-dienyl]benzene
SMILES C(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1

Specifications

Melting Point 151°C to 154°C
Boiling Point 350°C (720 mmHg)
Odor Odorless
Quantity 5 g
Beilstein 1905937
Merck Index 14,3320
Solubility Information Slightly soluble in water.
Formula Weight 206.29
Percent Purity ≥98%
Chemical Name or Material trans,trans-1,4-Diphenyl-1,3-butadiene
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-952-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only