missing translation for 'onlineSavingsMsg'
Learn More

trans-p-Coumaric acid, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014350
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014350 Supplier Merck Emd Millipore Supplier No. 5582350MG
Only null left

trans-p-coumaric acid is a constituent of cutin. It is also found in olive processing and wine distillery effluents.

Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 501-98-4
Physical Form Neat
Linear Formula HOC6H4 CH=CHCO2 H
MDL Number MFCD00004399
Melting Point 214°C
Synonym p-Coumaric acid; trans-4-Hydroxycinnamic acid
Type Analytical Standard
Quantity 50 mg
Formula Weight 164.16
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less