missing translation for 'onlineSavingsMsg'
Learn More

trans-o-Coumaric Acid 98.0+%, TCI America™

Catalog No. p-7124393
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03945G 5 g
C039425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C03945G Supplier TCI America Supplier No. C03945G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 614-60-8
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00004379
InChI Key PMOWTIHVNWZYFI-AATRIKPKSA-N
Synonym 2-hydroxycinnamic acid, o-coumaric acid, trans-2-hydroxycinnamic acid, 2-coumaric acid, trans-o-hydroxycinnamic acid, trans-o-coumaric acid, 2-coumarate, e-o-hydroxycinnamic acid, 2-hydroxycinnamate, o-hydroxy-trans-cinnamic acid
PubChem CID 637540
ChEBI CHEBI:18125
IUPAC Name (2E)-3-(2-hydroxyphenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CC=C1O

Specifications

Melting Point 217°C
Color White-Yellow
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-o-Coumaric Acid
RTECSNumber GD9090000
TSCA Yes