accessibility menu, dialog, popup

UserName

CAS RN 614-60-8

IUPAC Name: (2E)-3-(2-hydroxyphenyl)prop-2-enoic acid
Synonyms: trans-2-hydroxycinnamic acid 2-hydroxycinnamate O-hydroxycinnamic acid O-hydroxy-trans-cinnamic acid trans-2-hydroxycinnamate
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: PMOWTIHVNWZYFI-AATRIKPKSA-N
SMILES: OC(=O)\C=C\C1=CC=CC=C1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)