missing translation for 'onlineSavingsMsg'
Learn More

trans-N-Fmoc-4-hydroxy-L-proline, 97%

Catalog No. AAH5274003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5274003 1 g
AAH5274006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5274003 Supplier Thermo Scientific Chemicals Supplier No. H5274003

CAS: 88050-17-3 | C20H19NO5 | 353.374 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88050-17-3
Molecular Formula C20H19NO5
Molecular Weight (g/mol) 353.374
MDL Number MFCD00151929
InChI Key GOUUPUICWUFXPM-XIKOKIGWSA-N
Synonym fmoc-hyp-oh, fmoc-l-hydroxyproline, fmoc-l-4-hydroxyproline, fmoc-trans-4-hydroxy-l-proline, n-fmoc-trans-4-hydroxy-l-proline, fmoc-l-hyp-oh, trans-n-fmoc-4-hydroxy-l-proline, 2s,4r-1-9h-fluoren-9-yl methoxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s,4r-1-9h-fluoren-9-ylmethoxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, n-alpha-9-fluorenylmethyloxycarbonyl-trans-l-hydroxyproline
PubChem CID 2756109
IUPAC Name (2S,4R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES C1C(CN(C1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)O

Specifications

Melting Point 189°C to 193°C
Quantity 1 g
Optical Rotation −60° (c=1 in Methanol)
Formula Weight 353.37
Percent Purity 97%
Chemical Name or Material trans-N-Fmoc-4-hydroxy-L-proline
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only