missing translation for 'onlineSavingsMsg'
Learn More

trans-N-Boc-4-cyano-L-proline methyl ester, 97%, Thermo Scientific Chemicals

Catalog No. p-7035031
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5273303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5273303 Supplier Thermo Scientific Chemicals Supplier No. H5273303
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 194163-91-2
Molecular Formula C12H18N2O4
Molecular Weight (g/mol) 254.29
MDL Number MFCD03094915
InChI Key GHLKJIQVORAVHE-UHFFFAOYNA-N
Synonym boc-trans-4-cyano-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, n-boc-trans-4-cyano-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4r-4-cyanopyrrolidine-1,2-dicarboxylate, n-tert-butoxycarbonyl-trans-4-cyano-l-proline methyl ester, 4r-1-tert-butyloxycarbonyl-4-cyano-l-proline methyl ester, 2s,4r-1-tert-butoxycarbonyl-4-cyano-2-methylpyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid,4-cyano-,1-1,1-dimethylethyl 2-methyl ester, 2s,4r, 1,2-pyrrolidinedicarboxylic acid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4r, 1,2-pyrrolidinedicarboxylicacid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4r
PubChem CID 17998966
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4R)-4-cyanopyrrolidine-1,2-dicarboxylate
SMILES COC(=O)C1CC(CN1C(=O)OC(C)(C)C)C#N

Specifications

Quantity 1 g
Formula Weight 254.29
Percent Purity 97%
Chemical Name or Material trans-N-Boc-4-cyano-L-proline methyl ester
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only