missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-N-Boc-4-cyano-L-proline, 97%
CAS: 273221-94-6 | C11H16N2O4 | 240.259 g/mol
$624.92 - $1867.70
Chemical Identifiers
| CAS | 273221-94-6 |
|---|---|
| Molecular Formula | C11H16N2O4 |
| Molecular Weight (g/mol) | 240.259 |
| MDL Number | MFCD01860670 |
| InChI Key | MIVXQYMYEMUMKK-YUMQZZPRSA-N |
| Synonym | 2s,4r-1-tert-butoxycarbonyl-4-cyanopyrrolidine-2-carboxylic acid, n-boc-trans-4-cyano-l-proline, trans-n-boc-4-cyano-l-proline, 4-cyano-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, boc-trans-4-cyano-l-proline, trans-n-boc-4-cyano-l-proline 250mg, 4r-1-tert-butoxycarbonyl-4-cyano-l-proline |
| PubChem CID | 17750460 |
| IUPAC Name | (2S,4R)-4-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
| SMILES | CC(C)(C)OC(=O)N1CC(CC1C(=O)O)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH52563MD
|
Thermo Scientific Chemicals
H52563MD |
250 mg |
Each for $624.92
|
|
|||||
|
AAH5256303
|
Thermo Scientific Chemicals
H5256303 |
1 g |
Each for $1,867.70
|
|
|||||
Description
trans-N-Boc-4-cyano-L-proline is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-N-Boc-4-cyano-L-proline is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.
Chemical Identifiers
| 273221-94-6 | |
| 240.259 | |
| MIVXQYMYEMUMKK-YUMQZZPRSA-N | |
| 17750460 | |
| CC(C)(C)OC(=O)N1CC(CC1C(=O)O)C#N |
| C11H16N2O4 | |
| MFCD01860670 | |
| 2s,4r-1-tert-butoxycarbonyl-4-cyanopyrrolidine-2-carboxylic acid, n-boc-trans-4-cyano-l-proline, trans-n-boc-4-cyano-l-proline, 4-cyano-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, boc-trans-4-cyano-l-proline, trans-n-boc-4-cyano-l-proline 250mg, 4r-1-tert-butoxycarbonyl-4-cyano-l-proline | |
| (2S,4R)-4-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
Specifications
| 273221-94-6 | |
| MFCD01860670 | |
| 2s,4r-1-tert-butoxycarbonyl-4-cyanopyrrolidine-2-carboxylic acid, n-boc-trans-4-cyano-l-proline, trans-n-boc-4-cyano-l-proline, 4-cyano-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, boc-trans-4-cyano-l-proline, trans-n-boc-4-cyano-l-proline 250mg, 4r-1-tert-butoxycarbonyl-4-cyano-l-proline | |
| MIVXQYMYEMUMKK-YUMQZZPRSA-N | |
| (2S,4R)-4-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid | |
| 17750460 | |
| 97% |
| C11H16N2O4 | |
| 250 mg | |
| Slightly soluble in water. | |
| CC(C)(C)OC(=O)N1CC(CC1C(=O)O)C#N | |
| 240.259 | |
| 240.26 | |
| trans-N-Boc-4-cyano-L-proline |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only