CAS RN 273221-94-6
IUPAC Name:
(2S,4R)-1-[(tert-butoxy)carbonyl]-4-cyanopyrrolidine-2-carboxylic acid
Synonyms:
(2S,4R)-1-(tert-butoxycarbonyl)-4-cyanopyrrolidine-2-carboxylic acid
Molecular Weight (g/mol):
240.26
Molecular Formula:
C11H16N2O4
InChi Key:
MIVXQYMYEMUMKK-YUMQZZPRSA-N
SMILES:
CC(C)(C)OC(=O)N1C[C@@H](C[C@H]1C(O)=O)C#N
1
–
1
of
1
results
1
Promotions Available
trans-N-Boc-4-cyano-L-proline, 97%
trans-N-Boc-4-cyano-L-proline, 97%
CAS: 273221-94-6 Molecular Formula: C11H16N2O4 Molecular Weight (g/mol): 240.259 MDL Number: MFCD01860670 InChI Key: MIVXQYMYEMUMKK-YUMQZZPRSA-N Synonym: 2s,4r-1-tert-butoxycarbonyl-4-cyanopyrrolidine-2-carboxylic acid,n-boc-trans-4-cyano-l-proline,trans-n-boc-4-cyano-l-proline,4-cyano-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester,boc-trans-4-cyano-l-proline,trans-n-boc-4-cyano-l-proline 250mg,4r-1-tert-butoxycarbonyl-4-cyano-l-proline PubChem CID: 17750460 IUPAC Name: (2S,4R)-4-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid SMILES: CC(C)(C)OC(=O)N1CC(CC1C(=O)O)C#N