missing translation for 'onlineSavingsMsg'
Learn More

trans-N-Boc-4-cyano-L-proline, 97%

Catalog No. AAH52563MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52563MD 250 mg
AAH5256303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52563MD Supplier Thermo Scientific Chemicals Supplier No. H52563MD

missing translation for 'validMainframeMsg'

CAS: 273221-94-6 | C11H16N2O4 | 240.259 g/mol

trans-N-Boc-4-cyano-L-proline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-N-Boc-4-cyano-L-proline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.

Chemical Identifiers

CAS 273221-94-6
Molecular Formula C11H16N2O4
Molecular Weight (g/mol) 240.259
MDL Number MFCD01860670
InChI Key MIVXQYMYEMUMKK-YUMQZZPRSA-N
Synonym 2s,4r-1-tert-butoxycarbonyl-4-cyanopyrrolidine-2-carboxylic acid, n-boc-trans-4-cyano-l-proline, trans-n-boc-4-cyano-l-proline, 4-cyano-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, boc-trans-4-cyano-l-proline, trans-n-boc-4-cyano-l-proline 250mg, 4r-1-tert-butoxycarbonyl-4-cyano-l-proline
PubChem CID 17750460
IUPAC Name (2S,4R)-4-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)C#N

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 240.26
Percent Purity 97%
Chemical Name or Material trans-N-Boc-4-cyano-L-proline
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only