missing translation for 'onlineSavingsMsg'
Learn More

trans-Methyl crotonate, 96%

Catalog No. AC160841000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160841000 100 g Glass bottle
AC160845000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160841000 Supplier Thermo Scientific Chemicals Supplier No. 160841000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 623-43-8 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 623-43-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009287
InChI Key MCVVUJPXSBQTRZ-ONEGZZNKSA-N
Synonym methyl crotonate, e-methyl but-2-enoate, trans-methyl crotonate, methyl e-crotonate, methyl 2-butenoate, methyl trans-crotonate, 2-butenoic acid, methyl ester, 2e, trans-2-butenoic acid methyl ester, methyl trans-2-butenoate, e-crotonic acid methyl ester
PubChem CID 638132
SMILES COC(=O)\C=C\C

Specifications

Color Colorless
Density 0.9400g/mL
Boiling Point 120°C
Flash Point -1°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHCO2CH3
Refractive Index 1.4230 to 1.4250
Quantity 100 g
Beilstein 02, 410
Specific Gravity 0.94
Merck Index 10, 2586
Solubility Information Solubility in water: insoluble
IUPAC Name methyl (2E)-but-2-enoate
Formula Weight 100.12
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material trans-Methyl crotonate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
Wash f
EINECSNumber 210-793-7
RTECSNumber GQ5710000
TSCA TSCA

RUO – Research Use Only