accessibility menu, dialog, popup

UserName

CAS RN 623-43-8

IUPAC Name: methyl (2E)-but-2-enoate
Synonyms: methyl (2E)-but-2-enoate
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: MCVVUJPXSBQTRZ-ONEGZZNKSA-N
SMILES: COC(=O)\C=C\C
Boiling Point 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)