missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals trans-L-3-Hydroxyproline
$246.74 - $1078.81
Chemical Identifiers
| CAS | 4298-08-2 |
|---|---|
| Molecular Formula | C5H9NO3 |
| Molecular Weight (g/mol) | 131.13 |
| MDL Number | MFCD00216471 |
| InChI Key | BJBUEDPLEOHJGE-IMJSIDKUSA-N |
| IUPAC Name | (2S,3S)-3-hydroxypyrrolidine-2-carboxylic acid |
| SMILES | O[C@H]1CCN[C@@H]1C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC297791000
|
Thermo Scientific Chemicals
297791000 |
100 mg |
Each for $246.74
|
|
|||||
|
AC297795000
|
Thermo Scientific Chemicals
297795000 |
500 mg |
Each for $1,078.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4298-08-2 | |
| 131.13 | |
| BJBUEDPLEOHJGE-IMJSIDKUSA-N | |
| O[C@H]1CCN[C@@H]1C(O)=O |
| C5H9NO3 | |
| MFCD00216471 | |
| (2S,3S)-3-hydroxypyrrolidine-2-carboxylic acid |
Specifications
| 235.0°C | |
| C5H9NO3 | |
| -17.50 | |
| O[C@H]1CCN[C@@H]1C(O)=O | |
| 100 mg | |
| 131.13 |
| 4298-08-2 | |
| MFCD00216471 | |
| BJBUEDPLEOHJGE-IMJSIDKUSA-N | |
| (2S,3S)-3-hydroxypyrrolidine-2-carboxylic acid | |
| 131.13 | |
| trans-L-3-Hydroxyproline |
RUO – Research Use Only