CAS RN 4298-08-2
IUPAC Name:
(2S,3S)-3-hydroxypyrrolidine-2-carboxylic acid
Synonyms:
trans-3-hydroxy-L-proline L-threo-3-hydroxyproline
Molecular Weight (g/mol):
131.13
Molecular Formula:
C5H9NO3
InChi Key:
BJBUEDPLEOHJGE-IMJSIDKUSA-N
SMILES:
O[C@H]1CCN[C@@H]1C(O)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals trans-L-3-Hydroxyproline
CAS: 4298-08-2 Molecular Formula: C5H9NO3 Molecular Weight (g/mol): 131.13 MDL Number: MFCD00216471 InChI Key: BJBUEDPLEOHJGE-IMJSIDKUSA-N IUPAC Name: (2S,3S)-3-hydroxypyrrolidine-2-carboxylic acid SMILES: O[C@H]1CCN[C@@H]1C(O)=O