missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals trans-L-3-Hydroxyproline

Catalog No. AC297791000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC297791000 100 mg
AC297795000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC297791000 Supplier Thermo Scientific Chemicals Supplier No. 297791000
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4298-08-2
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00216471
InChI Key BJBUEDPLEOHJGE-IMJSIDKUSA-N
IUPAC Name (2S,3S)-3-hydroxypyrrolidine-2-carboxylic acid
SMILES O[C@H]1CCN[C@@H]1C(O)=O

Specifications

Melting Point 235.0°C
Specific Rotation -17.50
Quantity 100 mg
Formula Weight 131.13
Chemical Name or Material trans-L-3-Hydroxyproline

RUO – Research Use Only