missing translation for 'onlineSavingsMsg'
Learn More

trans-Hydrindane 98.0+%, TCI America™

Catalog No. H05051ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05051ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H05051ML Supplier TCI America Supplier No. H05051ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3296-50-2
Molecular Formula C9H16
Molecular Weight (g/mol) 124.23
MDL Number MFCD00085358,MFCD00085357
InChI Key BNRNAKTVFSZAFA-UHFFFAOYNA-N
Synonym trans-Bicyclo[4.3.0]nonane, trans-Hexahydroindane, trans-Octahydroindene
PubChem CID 643588
ChEBI CHEBI:49308
IUPAC Name octahydro-1H-indene
SMILES C1CC2CCCCC2C1

Specifications

Color Colorless
Boiling Point 159°C
Quantity 1 mL
UN Number 3295
Formula Weight 124.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-Hydrindane
EINECSNumber (4)-1970
TSCA No