accessibility menu, dialog, popup

UserName

CAS RN 3296-50-2

IUPAC Name: octahydro-1H-indene
Synonyms: hydrindane trans-bicylco[4.3.0]nonane hydrindan
Molecular Weight (g/mol): 124.23
Molecular Formula: C9H16
InChi Key: BNRNAKTVFSZAFA-UHFFFAOYNA-N
SMILES: C1CC2CCCCC2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results