missing translation for 'onlineSavingsMsg'
Learn More

trans-Ethyl crotonate, 96%

Catalog No. AC161381000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161381000 100 g Glass bottle
AC161385000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161381000 Supplier Thermo Scientific Chemicals Supplier No. 161381000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 623-70-1 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 623-70-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00009289
InChI Key ZFDIRQKJPRINOQ-HWKANZROSA-N
Synonym ethyl crotonate, ethyl trans-crotonate, ethyl e-but-2-enoate, ethyl trans-2-butenoate, crotonic acid, ethyl ester, 2-butenoic acid, ethyl ester, 2e, ethyl e-2-butenoate, 2-butenoic acid, ethyl ester, ethyl 2e-but-2-enoate, crotonic acid, ethyl ester, e
PubChem CID 429065
IUPAC Name ethyl (E)-but-2-enoate
SMILES CCOC(=O)\C=C\C

Specifications

Acidity 1mg KOH/g max.
Color Colorless
Density 0.9180g/mL
Boiling Point 142°C to 143°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4232 to 1.4272
Linear Formula CH3CH=CHCO2C2H5
Quantity 100 g
Beilstein 02, 411
Specific Gravity 0.918
Merck Index 10, 2586
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 114.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material trans-Ethyl crotonate, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-808-7
RTECSNumber GQ3500000
TSCA TSCA

RUO – Research Use Only