accessibility menu, dialog, popup

UserName

CH3OOH3C(E)

CAS RN 623-70-1

IUPAC Name: ethyl (2E)-but-2-enoate
Synonyms: ethyl crotonate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: ZFDIRQKJPRINOQ-HWKANZROSA-N
SMILES: CCOC(=O)\C=C\C
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)