missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Diethyl caronate, 90%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 88% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4425 to 1.4445 |
| Quantity | 1 g |
| Formula Weight | 214.26 |
| Percent Purity | 90% |
| Physical Form | Liquid |
| Chemical Name or Material | trans-Diethyl caronate |
RUO – Research Use Only