accessibility menu, dialog, popup

UserName

CH3OO(S)(S)OOCH3H3CCH3

CAS RN 67945-22-6

IUPAC Name: 1,2-diethyl (1S,2S)-3,3-dimethylcyclopropane-1,2-dicarboxylate
Synonyms: 1,2-diethyl (1S,2S)-3,3-dimethylcyclopropane-1,2-dicarboxylate
Molecular Weight (g/mol): 214.26
Molecular Formula: C11H18O4
InChi Key: MPRMNWDIYNBXRC-HTQZYQBOSA-N
SMILES: CCOC(=O)[C@H]1[C@H](C(=O)OCC)C1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results