missing translation for 'onlineSavingsMsg'
Learn More

trans-Dibromobis(triphenylphosphine)palladium(II), Pd 13.4%, Thermo Scientific Chemicals

Catalog No. p-7049279
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1370404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1370404 Supplier Thermo Scientific Chemicals Supplier No. 01370404

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrogenation

Hydrogenation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22180-53-6
Molecular Formula C36H30Br2P2Pd
Molecular Weight (g/mol) 790.81
MDL Number MFCD00134175
InChI Key MCSDDEAMPOYJJI-UHFFFAOYSA-L
Synonym trans-dibromobis triphenylphosphine palladium ii, palladium bis triphenylphosphine dibromide, dibromobis triphenylphosphine palladium ii, trans-dibromobis triphenylphosphine-palladium ii, palladium 2+ bis triphenylphosphine dibromide
PubChem CID 11104744
IUPAC Name dibromopalladium;triphenylphosphane
SMILES [Br-].[Br-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 2 g
Sensitivity Hygroscopic
Solubility Information Soluble in chloroform, toluene and benzene
Formula Weight 790.8
Concentration or Composition (by Analyte or Components) Pd 13.4%
Physical Form Powder
Chemical Name or Material trans-Dibromobis(triphenylphosphine)palladium(II)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only