missing translation for 'onlineSavingsMsg'
Learn More

trans-Decahydroquinoline 98.0+%, TCI America™

Catalog No. D29365G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29365G Supplier TCI America Supplier No. D29365G
Only null left

Chemical Identifiers

CAS 767-92-0
Molecular Formula C9H17N
Molecular Weight (g/mol) 139.242
MDL Number MFCD00137755
InChI Key POTIYWUALSJREP-DTWKUNHWSA-N
PubChem CID 6994352
IUPAC Name (4aS,8aR)-1,2,3,4,4a,5,6,7,8,8a-decahydroquinoline
SMILES C1CCC2C(C1)CCCN2

Specifications

Melting Point 48°C
Color White-Yellow
Boiling Point 203°C
Quantity 5 g
Formula Weight 139.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-Decahydroquinoline