accessibility menu, dialog, popup

UserName

CAS RN 767-92-0

(S)HN(R)

CAS RN 767-92-0

IUPAC Name: (4aR,8aS)-decahydroquinoline
Synonyms: (4aR,8aS)-decahydroquinoline
Molecular Weight (g/mol): 139.24
Molecular Formula: C9H17N
InChi Key: POTIYWUALSJREP-BDAKNGLRSA-N
SMILES: C1CC[C@@H]2NCCC[C@H]2C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results