missing translation for 'onlineSavingsMsg'
Learn More

trans-Crotononitrile (contains ca. 20% cis- isomer) 75.0+%, TCI America™

Catalog No. C313225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C313225ML 25 mL
C3132100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C313225ML Supplier TCI America Supplier No. C313225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 627-26-9
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.09
MDL Number MFCD00001935
InChI Key NKKMVIVFRUYPLQ-NSCUHMNNSA-N
Synonym trans-2-Butenenitrile
PubChem CID 637921
IUPAC Name (2E)-but-2-enenitrile
SMILES C\C=C\C#N

Specifications

Color Yellow
Quantity 25 mL
UN Number 1993
Formula Weight 67.09
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-Crotononitrile (contains ca. 20% cis- isomer)
EINECSNumber (2)-3313
RTECSNumber EM8020000
TSCA No