accessibility menu, dialog, popup

UserName

CH3N(E)

CAS RN 627-26-9

IUPAC Name: (2E)-but-2-enenitrile
Synonyms: crotononitrile
Molecular Weight (g/mol): 67.09
Molecular Formula: C4H5N
InChi Key: NKKMVIVFRUYPLQ-NSCUHMNNSA-N
SMILES: C\C=C\C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results