missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-beta-Styrylboronic acid pinacol ester, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 27°C to 33°C |
| Boiling Point | 128°C (4 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.529 |
| Quantity | 250 mg |
| Beilstein | 8617179 |
| IUPAC Name | 4,4,5,5-tetramethyl-2-[(E)-2-phenylethenyl]-1,3,2-dioxaborolane |
| Formula Weight | 230.12 |
| Percent Purity | 99% |
| Chemical Name or Material | trans-beta-Styrylboronic acid pinacol ester |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only