missing translation for 'onlineSavingsMsg'
Learn More

trans-beta-Nitrostyrene 98.0+%, TCI America™

Catalog No. p-7134038
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
N09415G 5 g
N094125G 25 g
2 options

Catalog No. N09415G

Supplier: TCI America N09415G

Only null left

Chemical Identifiers

CAS 5153-67-3
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00007402
InChI Key PIAOLBVUVDXHHL-VOTSOKGWSA-N
Synonym trans-beta-nitrostyrene, 2-nitrovinyl benzene, e-2-nitrovinyl benzene, beta-nitrostyrene, 2-nitro-1-phenylethylene, e-beta-nitrostyrene, 2-nitroethenyl benzene, 1-phenyl-2-nitroethene, beta-nitrosytrene, benzene, 2-nitroethenyl
PubChem CID 5284459
IUPAC Name [(1E)-2-nitroethenyl]benzene
SMILES [O-][N+](=O)\C=C\C1=CC=CC=C1

Specifications

Melting Point 57°C
Color Yellow
Boiling Point 260°C
Quantity 5 g
Formula Weight 149.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-beta-Nitrostyrene
EINECSNumber (3)-0388
RTECSNumber WL5460000
TSCA No
Recommended Storage Refrigerator