missing translation for 'onlineSavingsMsg'
Learn More

trans-beta-Nitrostyrene, 97+%

Catalog No. p-7046452
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1436706 5 g
AAL1436714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1436706 Supplier Thermo Scientific Chemicals Supplier No. L1436706
Only null left

CAS: 5153-67-3 | C8H7NO2 | 149.15 g/mol

trans-beta-Nitrostyrene is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-beta-Nitrostyrene is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.

Chemical Identifiers

CAS 5153-67-3
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00007402
InChI Key PIAOLBVUVDXHHL-VOTSOKGWSA-N
Synonym trans-beta-nitrostyrene, 2-nitrovinyl benzene, e-2-nitrovinyl benzene, beta-nitrostyrene, 2-nitro-1-phenylethylene, e-beta-nitrostyrene, 2-nitroethenyl benzene, 1-phenyl-2-nitroethene, beta-nitrosytrene, benzene, 2-nitroethenyl
PubChem CID 5284459
IUPAC Name [(E)-2-nitroethenyl]benzene
SMILES [O-][N+](=O)\C=C\C1=CC=CC=C1

Specifications

Melting Point 55°C to 58°C
Boiling Point 250°C to 260°C
Quantity 5 g
Beilstein 1210066
Solubility Information Insoluble in water.
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material trans-beta-Nitrostyrene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-066-0
RTECSNumber WL5460000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only