missing translation for 'onlineSavingsMsg'
Learn More

trans-Benzalacetone 98.0+%, TCI America™

Catalog No. P0163500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0163500G 500 g
P016325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0163500G Supplier TCI America Supplier No. P0163500G

Chemical Identifiers

CAS 1896-62-4
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
MDL Number MFCD00008779
InChI Key BWHOZHOGCMHOBV-BQYQJAHWSA-N
Synonym benzalacetone, benzylideneacetone, methyl styryl ketone, 4-phenylbut-3-en-2-one, acetocinnamone, 4-phenyl-3-buten-2-one, e-4-phenylbut-3-en-2-one, benzylidene acetone, trans-benzalacetone, benzalaceton
PubChem CID 637759
ChEBI CHEBI:78399
IUPAC Name (E)-4-phenylbut-3-en-2-one
SMILES CC(=O)C=CC1=CC=CC=C1

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 500 g
UN Number 3077
Formula Weight 146.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material trans-Benzalacetone
EINECSNumber (3)-1232
RTECSNumber EN0330050
TSCA Yes