missing translation for 'onlineSavingsMsg'
Learn More

trans-Anethole, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014166
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014166 1 mL
111014167 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014166 Supplier Merck Emd Millipore Supplier No. 103681ML

trans-Anethole is a naturally occuring flavouring agent. It has insecticidal, larvicidal, and antimicrobial properties.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 4180-23-8
Solution Contains ˜0.05% 4-tert-butylcatechol as stabilizer
Density 0.988 g/mL (at 25°C)
Flash Point 91°C
Physical Form Neat
Linear Formula CH3 CH=CHC6H4 OCH3
MDL Number MFCD00009284
Melting Point 20°C to 21°C
Boiling Point 234°C to 237°C
Synonym 4-Propenylanisole; trans-1-Methoxy-4-(1-propenyl)benzene
Type Analytical Standard
Quantity 1 mL
Formula Weight 148.20
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less