missing translation for 'onlineSavingsMsg'
Learn More

trans-4-tert-Butylcyclohexanol 95.0+%, TCI America™

Catalog No. B12711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12711G 1 g
B12715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12711G Supplier TCI America Supplier No. B12711G

Chemical Identifiers

CAS 21862-63-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00064952
InChI Key CCOQPGVQAWPUPE-UHFFFAOYSA-N
PubChem CID 7391
IUPAC Name 4-tert-butylcyclohexan-1-ol
SMILES CC(C)(C)C1CCC(CC1)O

Specifications

Melting Point 80°C
Color White
Quantity 1 g
Formula Weight 156.27
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-tert-Butylcyclohexanol