accessibility menu, dialog, popup

UserName

CH3H3CCH3OH

CAS RN 21862-63-5

IUPAC Name: 4-tert-butylcyclohexan-1-ol
Synonyms: 4-T-butylcyclohexanol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: CCOQPGVQAWPUPE-UHFFFAOYSA-N
SMILES: CC(C)(C)C1CCC(O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results