missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America B325025G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 53292-89-0 | |
| 243.30 | |
| KXMRDHPZQHAXML-UHFFFAOYSA-N | |
| 2755996 | |
| CC(C)(C)OC(=O)NC1CCC(CC1)C(O)=O |
| C12H21NO4 | |
| MFCD01862293,MFCD03453262,MFCD01862294 | |
| trans-4-boc-amino cyclohexanecarboxylic acid, cis-4-boc-amino cyclohexanecarboxylic acid, 4-boc-amino cyclohexanecarboxylic acid, cis-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, trans-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-4-aminocyclohexanecarboxylic acid, boc-1,4-trans-achc-oh, cis-4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonylamino cyclohexanecarboxylic acid | |
| 4-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylic acid |
Specifications
| 183°C | |
| 53292-89-0 | |
| C12H21NO4 | |
| trans-4-boc-amino cyclohexanecarboxylic acid, cis-4-boc-amino cyclohexanecarboxylic acid, 4-boc-amino cyclohexanecarboxylic acid, cis-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, trans-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-4-aminocyclohexanecarboxylic acid, boc-1,4-trans-achc-oh, cis-4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonylamino cyclohexanecarboxylic acid | |
| CC(C)(C)OC(=O)NC1CCC(CC1)C(O)=O | |
| 25 g | |
| 2755996 | |
| ≥98.0% (GC,T) |
| trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid | |
| White-Yellow | |
| MFCD01862293,MFCD03453262,MFCD01862294 | |
| KXMRDHPZQHAXML-UHFFFAOYSA-N | |
| 4-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylic acid | |
| 243.30 | |
| 243.30 | |
| Crystalline Powder |
Safety and Handling
TSCA : No