missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7122835
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32505G 5 g
B325025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32505G Supplier TCI America Supplier No. B32505G

Chemical Identifiers

CAS 53292-89-0
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.30
MDL Number MFCD01862293,MFCD03453262,MFCD01862294
InChI Key KXMRDHPZQHAXML-UHFFFAOYSA-N
Synonym trans-4-boc-amino cyclohexanecarboxylic acid, cis-4-boc-amino cyclohexanecarboxylic acid, 4-boc-amino cyclohexanecarboxylic acid, cis-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, trans-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-4-aminocyclohexanecarboxylic acid, boc-1,4-trans-achc-oh, cis-4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonylamino cyclohexanecarboxylic acid
PubChem CID 2755996
IUPAC Name 4-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C(O)=O

Specifications

Melting Point 183°C
Color White-Yellow
Quantity 5 g
Formula Weight 243.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid