missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Propylcyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7134862
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09435G 5 g
P094325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09435G Supplier TCI America Supplier No. P09435G

Chemical Identifiers

CAS 38289-27-9
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00059559
InChI Key QCNUKEGGHOLBES-UHFFFAOYSA-N
Synonym trans-4-propylcyclohexanecarboxylic acid, 4-propylcyclohexanecarboxylic acid, trans-4-n-propylcyclohexanecarboxylic acid, 1s,4r-4-propylcyclohexane-1-carboxylic acid, propylcyclohexanecarboxylicacid, trans-4-propylcyclohexylcarboxylic acid, 4-trans-propyl cyclohexane carboxylic acid, trans-4-n-propyl-cyclohexanecarboxylicacid, trans-4-n-propylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-propyl-, trans
PubChem CID 1490324
IUPAC Name 4-propylcyclohexane-1-carboxylic acid
SMILES CCCC1CCC(CC1)C(=O)O

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 170.25
Percent Purity ≥98.0% (T,GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Propylcyclohexanecarboxylic Acid
EINECSNumber (3)-3723
TSCA No