CAS RN 38289-27-9
IUPAC Name:
4-propylcyclohexane-1-carboxylic acid
Synonyms:
4-propylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
170.25
Molecular Formula:
C10H18O2
InChi Key:
QCNUKEGGHOLBES-UHFFFAOYSA-N
SMILES:
CCCC1CCC(CC1)C(O)=O
1
–
1
of
1
results
1
trans-4-Propylcyclohexanecarboxylic Acid 98.0+%, TCI America™
CAS: 38289-27-9 Molecular Formula: C10H18O2 Molecular Weight (g/mol): 170.252 MDL Number: MFCD00059559 InChI Key: QCNUKEGGHOLBES-UHFFFAOYSA-N Synonym: trans-4-propylcyclohexanecarboxylic acid,4-propylcyclohexanecarboxylic acid,trans-4-n-propylcyclohexanecarboxylic acid,1s,4r-4-propylcyclohexane-1-carboxylic acid,propylcyclohexanecarboxylicacid,trans-4-propylcyclohexylcarboxylic acid,4-trans-propyl cyclohexane carboxylic acid,trans-4-n-propyl-cyclohexanecarboxylicacid,trans-4-n-propylcyclohexane carboxylic acid,cyclohexanecarboxylic acid, 4-propyl-, trans PubChem CID: 1490324 IUPAC Name: 4-propylcyclohexane-1-carboxylic acid SMILES: CCCC1CCC(CC1)C(=O)O